Learn More
Ansell AlphaTec™ 2000 STANDARD Jacket & Trouser Set Bound
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Dimensione della confezione:
pezzi 40
Descrizione
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifica
Specifica
| Size | X-Large |
| Sesso | Unisex |
| Materiale | Microporous Non-woven Polyethylene Laminate |
| Colore | White |
| Tipo di chiusura | Zipper |
| Tipo di giuntura | Bound |
| Certifications/Compliance (Europe) | Partial Body Protection Only, EN 13034:2005 + A1:2009, DIN 32781:2010, EN 14126:2003, ASTM F1671, EN ISO 13688:2013 |
| Product Type | Chemical-resistant Jacket |
| Stile | W20B219 |
| Da utilizzare con (applicazione) | Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |