Learn More
Ansell AlphaTec™ 2000 STANDARD Overboots Bound
Codice prodotto. p-7228377
Click to view available options
Stile:
W20B406
W20B407
Dimensione della confezione:
150 paia
200 paia
Codice prodotto. 16804800
Marca: Ansell WH20B0040600
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Comfortable overboots featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifica
Size Standard
Materiale Non-woven Polypropylene Laminate
Latex Free Yes
Colore White
Monouso Limited-use
Sterilità Non-sterile
Tipo di chiusura Elastic Top with Ankle Ties
Tipo di suola Standard
Tipo di giuntura Bound
Stile W20B406
Da utilizzare con (applicazione) Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services
Certifications/Compliance (Europe) EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013, Partial Body Protection Only
Peso (sistema metrico) 63 gsm
Resistenza Chemical-resistant
Product Type Controlled Environments Ankle High Shoe Covers