Learn More
Ansell AlphaTec™ 2000 STANDARD Trousers Bound
Click to view available options
Size:
X-Large
2X-Large
Dimensione della confezione:
pezzi 80
Descrizione
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifica
Specifica
| Size | X-Large |
| Sesso | Unisex |
| Product Type | Chemical-resistant Trousers |
| Materiale | Microporous Non-woven Polyethylene Laminate |
| Colore | White |
| Tipo di giuntura | Bound |
| Certifications/Compliance (Europe) | Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013 |
| Stile | W20B301 |
| Da utilizzare con (applicazione) | Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |