Learn More
Ansell AlphaTec™ 2500 STANDARD
Codice prodotto. 16874860
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Dimensione della confezione:
pezzi 20
pezzi 25
pezzi 30
Codice prodotto. 16874860
Marca: Ansell WH25W0012205
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Chemical protective suits with durable and breathable microporous laminate technology and Type 4/5/6 protection.
  • Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
  • Ultra low linting to help reduce the risk of cross contamination in critical areas
  • Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifica
Colore White
Da utilizzare con (applicazione) Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments
Hood Type Attached
Materiale Microporous Non-woven Polypropylene Laminate
Size X-Large
Product Type Ultrasonically Welded Coverall
Tipo di giuntura Welded
Stile W25W122
Peso (sistema metrico) 65 gsm