Learn More
Ansell AlphaTec™ 2500 STANDARD
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Dimensione della confezione:
pezzi 20
pezzi 25
pezzi 30
Descrizione
- Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
- Ultra low linting to help reduce the risk of cross contamination in critical areas
- Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
- Anti-static
Specifica
Specifica
| Colore | White |
| Da utilizzare con (applicazione) | Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments |
| Hood Type | Attached |
| Materiale | Microporous Non-woven Polypropylene Laminate |
| Size | 2X-Large |
| Product Type | Ultrasonically Welded Coverall |
| Tipo di giuntura | Welded |
| Stile | W25W122 |
| Peso (sistema metrico) | 65 gsm |