Learn More
Ansell AlphaTec 3000 AVANT AIRline
Codice prodotto. p-7228391
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Stile:
Y30T755
Y30T755D
Dimensione della confezione:
pezzi 4
Codice prodotto. 16814990
Marca: Ansell YE30T0075502
Encapsulated chemical protective suits for use with air-fed/supplied systems.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress.
TRUSTED_SUSTAINABILITY
Specifica
Colore Yellow
Da utilizzare con (applicazione) Transferring liquids and solids between vessels and tanks and process equipment, Opening and draining pumps valves or lines, Filling, blending, and charging of raw materials, Mixing or loading chemicals, Cleaning of plant and machines, Unexpected leakages spills or other releases, Loading and unloading trucks and vehicles, Product inspection
Hood Type Fully Encapsulated
Materiale Multi-layer Barrier Laminate Fabric
Size Small
Product Type Encapsulated chemical protective suits for use with air-fed/supplied systems
Tipo di giuntura Welded and Taped
Stile Y30T755
Peso (sistema metrico) 80 gsm