Learn More
Ansell™ AlphaTec™ 3000 AVANT AIRline Ultrasonically Welded and Taped
Codice prodotto. p-200004974
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 4
This item is not returnable. View return policy
Product Code. 16894970
Brand: Ansell™ YE30T0075402
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
AlphaTec™ 3000 AVANT AIRline Ultrasonically welded and taped is a fully encapsulated suit designed for use in combination with the AlphaTec™ AVANT STS V.1 airline regulator.
Fully encapsulated suit designed to be used with the permanently attached MICROCHEM™ AVANT STS V.1 airline regulator.
  • Features include air-channel ventilation system
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
TRUSTED_SUSTAINABILITY
Specifications
Certificazione/Conformità CE CATEGORY III (Type 3, EN 1073-1, EN 14594, EN 1149-5)
Colore Yellow
Da utilizzare con (applicazione) Transferring liquids and solids between vessels and tanks and process equipment Opening and draining pumps valves or lines Filling, blending and charging of raw materials Mixing or loading chemicals Cleaning of plant and machines Unexpected leakages spills or other releases Loading and unloading trucks and vehicles Product inspection
Hood Type Fully Encapsulated
Materiale Multi-layer Barrier Laminate Fabric
Size Small
Stile della caviglia Attached Sock Boots with Anti-slip PVC Sole
Tipo di chiusura Zipper
Stile polso Double cuffs and permanently attached chemical protective gloves
Monouso Reusable
Sesso Unisex
Stile collo Hood
N. di tasche None
Resistenza Chemical I Radioactive I Antistaic
Tipo di giuntura ULTRASONICALLY WELDED AND TAPED
Sterilità Non-sterile
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.