Learn More
Ansell AlphaTec™ 3000 AVANT2 AIRline
Codice prodotto. 17121075
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 4
Codice prodotto. 17121075
Marca: Ansell YE30T00075707G05
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Encapsulated chemical protective suits for use with air-fed/supplied systems.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Panoramic visor design ensures a good field of vision for the wearer.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow). 
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer.
TRUSTED_SUSTAINABILITY
Specifica
Colore Yellow
Da utilizzare con (applicazione) Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Opening and Draining Pumps Valves or Lines, Filling, Blending and Charging of Raw Materials, Mixing or Loading Chemicals, Cleaning of Plant and Machines, Unexpected Leakages Spills or Other Releases, Loading and Unloading Trucks and Vehicles, Product Inspection
Hood Type Fully Encapsulated
Materiale Multi-layer Barrier Laminate Fabric
Size 3X-Large
Product Type Encapsulated chemical protective suits for use with air-fed/supplied systems
Tipo di giuntura Welded and Taped
Stile Y300757D
Peso (sistema metrico) 80 gsm