Learn More
Ansell AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped - Model 756-G05
Codice prodotto. p-200080417
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 4
Codice prodotto. 17181075
Marca: Ansell YE30TA0075603G05
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped Chemical Protective Suit features a panoramic visor design that ensures an unobstructed field of vision.
  • Specialized protection: The AlphaTec 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 chemical protective suit has proven to protect wearers from over 200 chemicals.
  • Enhanced features: This full protective suit features a panoramic visor design that ensures an unobstructed field of vision
  • Genuine versatility: The AlphaTec 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 full chemical protection suit also has an HEPA filter, which provides secondary protection from airline contamination and an SMC silencer keeps noise in the suit below 70 dB (even at maximum airflow)
  • Considered aesthetics: In addition, the attached socks with elasticated boot overflap was designed to be worn inside chemical protective boots with the leg overflap worn outside, to reduce the potential for chemical ingress
DESIGN FEATURES:
  • Fully encapsulated suit designed for use in combination with the permanently attached AlphaTec™ AVANT STS V.1 airline regulator.
  • Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop, G01 AlphaTec™ 37-676 or G05 AlphaTec™ 29-500 gloves.
  • Attached boot ends with anti-slip PVC sole version also available and emergency rip cord.
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Colore Yellow
Da utilizzare con (applicazione) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes
Hood Type Fully Encapsulated
Materiale Multi-layer Barrier Laminate Fabric
Size Medium
Stile della caviglia Elasticated Boot Overflap
Tipo di colletto Encapsulated hood
Monouso Reusable
Sesso Unisex
Product Type Chemical Protective Suit
Classe di protezione CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Tipo di giuntura Welded and Taped
Sterilità Non-sterile