Learn More
Ansell AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped - Model 757 - G05
Codice prodotto. 17161095
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 4
Codice prodotto. 17161095
Marca: Ansell YE30TA0075703G05
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped chemical protective suit's HEPA filter offers secondary protection from airline contamination and a SMC silencer maintains the noise within a suit below 70 dB (even at maximum airflow).
  • Specialized protection: The AlphaTec™ 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 757 full body protection suit has been tested against 200 chemicals.
  • Genuine versatility: This chemical protective suit's HEPA filter offers secondary protection from airline contamination and a SMC silencer maintains the noise within a suit below 70 dB (even at maximum airflow)
  • Considered aesthetics: In addition, this protective suit has attached socks with an elasticated boot overflap and is designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
  • Reduced risks: The emergency ripcord feature of this full protective suit permits quick doffing of the suit in cases of emergency or undue distress to the wearer
DESIGN FEATURES:
  • Fully encapsulated suit designed for use in combination with the permanently attached AlphaTec™ AVANT STS V.1 airline regulator.
  • Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop, G01 Ansell Solvex (Nitrile) gloves, G02 Ansell Barrier gloves or G05 AlphaTec™ 29-500 gloves.
  • Attached socks with elasticated boot over-flap and emergency rip cord.
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Colore Yellow
Da utilizzare con (applicazione) Opening and draining pumps valves or lines, Filling, blending and charging of raw materials, Contaminated Environments, Chemical Handling, Transferring liquids and solids, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Loading and unloading of proces equipment
Hood Type Fully Encapsulated
Materiale Multi-layer Barrier Laminate Fabric
Size Medium
Stile della caviglia Attached Socks, Elasticated Boot Over Flap
Monouso Reusable
Da utilizzare con (apparecchiatura) Air-Fed/Supplied Systems
Sesso Unisex
Stile collo Hood
Product Type Encapsulated Chemical Protective Suit
Classe di protezione CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Tipo di giuntura Welded and Taped
Sterilità Non-sterile