Learn More
Ansell™ AlphaTec™ 4000 AVANT AIRline Ultrasonically Welded and Taped
Codice prodotto. p-200004970
Click to view available options
Size:
2X-Large
3X-Large
Large
Large/X-Large
Medium
Small
Dimensione della confezione:
pezzi 3
Codice prodotto. 17120735
Marca: Ansell™ GR40T0075402
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
AlphaTec™ 4000 AVANT AIRline Ultrasonically welded and taped are affixed by the wearer and detachable for reuse when appropriate. Air flow is adjustable from 340 L/min to 590 L/min at a working pressure of 3.5 to 5 bar.
Affixed by the wearer and detachable.
  • Air flow is adjustable from 340 L/min to 590 L/min at a working pressure of 3.5 to 5 bar.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached sock boots with anti-slip PVC sole - Designed to allow soft or protective safety shoes to be worn inside and offer limited resistance to slip
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità CE CATEGORY III (Type 3, EN 1073-1, EN 14594, EN 1149-5)
Colore Green
Da utilizzare con (applicazione) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes
Hood Type Fully Encapsulated
Materiale Multi-layer Barrier Laminate Fabric
Size Small
Stile della caviglia Attached Elastic Sock Boots with Anti-slip PVC Sole
Tipo di chiusura Zipper
Stile polso Elasticated double cuffs and permanently attached chemical protective gloves
Monouso Reusable
Sesso Unisex
Stile collo Fully encapsulated hood with visor
N. di tasche None
Classe di protezione CE CATEGORY III
Resistenza Chemical I Particle I Radioactive I ESD
Tipo di giuntura Welded and Taped
Sterilità Non-sterile