Descrizione
- Specialized protection: AlphaTec™ 4000 CFR Stitched and Taped - Model 111 chemical protective suit features multi-hazard solutions that offer a proven 8 hour barrier against chemicals such as non-flammable, flammable and aggressive solvents
- Improved durability: Complying with EN ISO 14116 and ASTM D6413 Flame Resistance*, this full protective suit shields the wearer from chemical sprays without compromising safety by limiting flame spread in the event of a flash fire
- Elevated comfort: The optimized body fit design of this protective suit offers improved wearer comfort and safety
- This full-chemical protection suit defends against infective agents, tested according to ASTM F1671 and hydrocarbons such as hexane, HF, oleum and many more
- *AlphaTec™ 4000 CFR should never be worn in isolation for flame retardant protection.
- Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above
- Polyolefin-based core barrier laminate, with FR non-woven inner and FR outer barrier film
Specifica
Specifica
| Product Type | Chemical-resistant Coveralls |
| Size | 5X-Large |
| Sesso | Unisex |
| Materiale | Polyolefin |
| Colore | Orange |
| Monouso | Single-use |
| Stile vita | Elastic at Back |
| Stile della caviglia | Elastic |
| Stile polso | Elastic |
| Tipo di cappa | Attached |
| Vedi altri risultati |