Learn More
Ansell AlphaTec™ 4000 Jacket
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 20
Descrizione
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect
- Silicone free
Specifica
Specifica
| Size | X-Large |
| Sesso | Unisex |
| Materiale | Multi-layer Barrier Laminate Fabric |
| Colore | Green |
| Tipo di chiusura | Dual Zipper |
| Tipo di giuntura | Welded and Taped |
| Certifications/Compliance (Europe) | Partial Body Protection Only, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 14126:2003 |
| Product Type | Chemical-resistant Jacket |
| Stile | G40T230 |
| Da utilizzare con (applicazione) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
| Vedi altri risultati |