Learn More
Ansell AlphaTec™ 58-335 Nitrile Gloves With AQUADRI™ Technology
Codice prodotto. p-6983007
Click to view available options
Size:
11
Dimensione della confezione:
12 paia
Codice prodotto. 15335171
Marca: Ansell 58335110
Solo null disponibili
Aggiungi al carrello
Edge
Aggiungi al carrello
Chemical gloves with new AQUADRI™ moisture management technology keeps hands dryer for significantly longer periods of time.
  • Nitrile film gives advanced chemical protection against incidental exposure to bases, oils, fuels, some solvents, greases and animal fats
  • Reversed-lozenge finish provides improved dry and wet grip, delivering optimal results in dry or wet conditions
  • High-performance nitrile compound offers snag, puncture and abrasion protection compared to rubber or neoprene
  • Foldable cuff gutter prevents dripping on the forearm
TRUSTED_SUSTAINABILITY
Specifica
ANSI Abrasion Rating ANSI 5
Certificazione/Conformità 21 CFR 170-199
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, Category III, EN 1149
Chlorinated Yes
Applicazioni Industrial
Colore Green
Finger Thickness (English) 0.030 in.
Finger Thickness (Metric) 0.75 mm
Hand Ambidestri
Latex Free
Materiale Nitrile
Palm Thickness (English) 0.0291 in.
Palm Thickness (Metric) 0.74 mm
Product Type Chemical-resistant Gloves
Size 11
Spessore (sistema imperiale) 31 mil
Spessore (sistema metrico) 0.78 mm
EN388 Standard EN 388:4101A, EN 388:4101
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPT
EN ISO 374 - 5 Bacteria, Fungi only
Da utilizzare con (applicazione) Automotive, chemical, machinery and equipment, metal fabrication
Stile polso guanto Guanto con manichetta
Grip Design Reversed Lozenge
Materiale rivestimento interno AQUADRI™ Technology
Rivestimento interno Lined
Modello 58-335 11 114023
Serie AlphaTec™ 58-335 AQUADRI™
Lunghezza (sistema imperiale) 15 in.
Lunghezza (sistema metrico) 38 cm