Learn More
Thermo Scientific Chemicals Aluminum oxide, for chromatography, acidic, Brockmann I, 50-200 μm, 90A
Codice prodotto. 16504204 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Click to view available options
Quantità:
1 kg
Dimensione della confezione:
1kg
Codice prodotto. 16504204
Marca: Thermo Scientific Chemicals 190400010
Richiesta di formato massivo o personalizzato
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 1 kg