Learn More
Thermo Scientific Chemicals Aluminum Dish, Flat Bottom & Straight Sides;Dia (mm), 76;Wall Ht (mm), 25
Click to view available options
Diametro (sistema metrico):
51 mm
76 mm
90 mm
126 mm
204 mm
Altezza (sistema metrico):
12.5 mm
25 mm
51 mm
N. per confezione:
2/Pk.
6/Pk.
Dimensione della confezione:
pezzi 2
pezzi 6
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Forma del fondo | Flat |
| Materiale | Aluminum |
| Forma | Flat Bottom and Straight Sides |
| Tipo | Dish |
| N. per confezione | 2/Pk. |
| Altezza (sistema metrico) | 25 mm |
| Diametro (sistema metrico) | 76 mm |