Learn More
Buerkle™ Aluminum Profisampler™
Sfoglia Tutto Bürkle Prodotti
Click to view available options
Materiale:
Aluminum
Dimensione della confezione:
Pezzo
Descrizione
The ProfiSampler aluminum set consists of a vacuum pump made of aluminum, with feet and venting valve and bottle thread GL 45, glass bottle 500mL, five suction tubes (black PA) 220cm long and electrically conductive, hose cutter, brass hose weight, 20 Seal-it bottle seals and an earthing cable.
- Stands on three foot supports for stability and safety
- Fill level control, automatic return flow and overfill protection through built-in venting valve.
- Available with suction hose in PE, PA and PTFE wherever the highest degree of purity is required
- Can be supplied with straight suction tubes for simpler sampling, including a transport case for convenient carriage and disposal of the suction tubes
- Suction volume 200mL, suction pressure 0.5 bar
- Suction height from liquid surface 5m
- Sample bottles available in different materials and sizes
Specifica
Specifica
| Materiale | Aluminum |
| Autoclavabile | Autoclavable |