Learn More
Cole-Parmer™ Aluminum Crimp-walled Weighing Dish
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Capacità (sistema metrico):
20 mL
Dimensione della confezione:
pezzi 1000
Descrizione
Use these weighing dishes for a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical and pharmaceutical. Aluminum Crimp-walled Weighing Dishes feature crimped sides for rigidity, sturdy gauge for heavy duty use.
- Dishes feature large tab for sample numbers
- Flat bottom to improve sample handling
- Sides are angled for easy separation
- No oil residue, no vinyl coating to create contamination problems.
- Smooth bottom
- Crimpled walled with side finger tab handle
- Disposable
Specifica
Specifica
| Forma del fondo | Flat |
| Capacità (sistema metrico) | 20 mL |
| Monouso | Single-use |
| Materiale | Aluminum |
| Forma | Round |
| Tipo | Weighing Dish |
| Profondità (sistema metrico) | 4.3 cm |
| Diametro (sistema imperiale) | 1.75 in. |
| Diametro (sistema metrico) | 1.3 cm |
Titolo del prodotto
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu