Learn More
Invitrogen™ Ambion™ RNase A (RPA grade) 1mg/mL
Codice prodotto. 10763267
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
Pezzo
Codice prodotto. 10763267
Marca: Invitrogen™ AM2272
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Endonuclease-free, exonuclease-free, protease-free
RPA Grade RNase A is intended for those who desire to use RNase A alone in ribonuclease protection assays instead of usual mixture of RNases A and T1. This product is equivalent to molecular biology grade RNase A from other suppliers.
  • Endonuclease that specifically cleaves 3' of U and C residues
  • Supplied in one tube containing 1mg (1mg/mL)
  • Free of DNase and nicking activities
  • Rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity and functionality is determined in a standard RPA III™ (Mfr. No. AM1414) reaction
DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification
Order Info
Shipping Conditions: Dry ice
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage -20°C
Quantità 1 mg