missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMBRA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-24723
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
AMBRA1 Polyclonal specifically detects AMBRA1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| AMBRA1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| activating molecule in BECN1-regulated autophagy protein 1, autophagy/beclin-1 regulator 1, DCAF3, DDB1 and CUL4 associated factor 3, FLJ20294, KIAA1736activating molecule in beclin-1-regulated autophagy, MGC33725, WD repeat domain 94, WDR94 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the antibody is 100% homologous in human, zebra fish, wild boar, duck billed platypus, rat, mouse, chimpanzee, opossum, macaques, monkey, dog, xenopus and chicken. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Q9C0C7 | |
| AMBRA1 | |
| A portion of amino acids 1-50 of human Ambra1 was used as the immunogen for the antibody. | |
| 0.1 mg | |
| Autophagy | |
| 55626 | |
| Human, Mouse, Rat, Bovine, Chicken | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto