Learn More
Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals
24.00€ - 183.00€
Identificatori chimici
CAS | 6009-70-7 |
---|---|
Formula molecolare | C2H10N2O5 |
Molecular Weight (g/mol) | 142.111 |
Numero MDL | MFCD00149694 |
InChI Key | MSMNVXKYCPHLLN-UHFFFAOYSA-N |
Sinonimo | ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate |
PubChem CID | 516808 |
IUPAC Name | diazanium;oxalate;hydrate |
SMILES | C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11493973
|
Thermo Scientific Alfa Aesar
A10263.22 |
100 g |
24.00€
100g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11403983
|
Thermo Scientific Alfa Aesar
A10263.36 |
500 g |
53.70€
500g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11413983
|
Thermo Scientific Alfa Aesar
A10263.0E |
2500 g |
183.00€
2500g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Incompatible with strong oxidizing agents and strong acids.
Identificatori chimici
6009-70-7 | |
142.111 | |
MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
516808 | |
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O |
C2H10N2O5 | |
MFCD00149694 | |
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
diazanium;oxalate;hydrate |
Specifica
Ammonium oxalate monohydrate | |
70°C (decomposition) | |
>99% | |
(COONH4)2·H2O | |
UN3288 | |
14,537 | |
Soluble in water. Slightly soluble in alcohol. | |
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O | |
142.111 | |
142.11 (124.10Anhydrous) |
6009-70-7 | |
1.5 g/mL | |
C2H10N2O5 | |
MFCD00149694 | |
3759830 | |
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
diazanium;oxalate;hydrate | |
516808 | |
98% |
Sicurezza e movimentazione
GHS H Statement
H302-H312
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312-H320-H335-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 238-135-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only