missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AMPH (Human) Recombinant Protein
Click to view available options
Quantità:
10 μg
25 μg
Dimensione della confezione:
10µg
25µg
Descrizione
- Sequence: MADIKTGIFAKNVQKRLNRAQEKVLQKLGKADETKDEQFEEYVQNFKRQEAEGTRLQRELRGYLAAIKGMQEASMKLTESLHEVYEPDWYGREDVKMVGEKCDVLWEDFHQKLVDGSLLTLDTYLGQFPDIKNRIAKRSRKLVDYDSARHHLEALQSSKRKDESRISKAEEEFQKAQKVFEEFNVDLQEELPSLWSRRVGFYVNTFKNVSSLEAKFHKEIAVLCHKLYEVMTKLGDQHADKAFTIQGAPSDSGPLRIAKTPSPPEEPSPLPSPTASPNHTLAPASPAPARPRSPSQTRKGPPVPPLPKVTPTKELQQENIISFFEDNFVPEISVTTPSQNEVPEVKKEETLLDLDFDPFKPEVTPAGSAGVTHSPMSQTLPWDLWTTSTDLVQPASGGSFNGFTQPQDTSLFTMQTDQSMICNLAESEQAPPTEPKAEEPLAAVTPAVGLDLGMDTRAEEPVEEAVIIPGADADAAVGTLVSAAEGAPGEEAEAETATVPAGEGVSLEEAKIGTETTEGAESAQPEAEELEATVPQEKVIPSVVIEPASNHEEEGENEITIGAEPKETTEDAAPPGPTSETPELATEQKPIQDPQPTPSAPAMGAADQLASAREASQELPPGFLYKVETLHDFEAANSDELTLQRGDVVLVVPSDSEADQDAGWLVGVKESDWLQYRDLATYKGLFPENFTRRLD
Specifica
Specifica
| Numero di accesso | AAH34376.1 |
| ID gene (immissione) | 273 |
| Nome | amphiphysin |
| Metodo di preparazione | Wheat germ expression system |
| Analisi di controllo qualità | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantità | 25 μg |
| Requisiti di stoccaggio | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Alias gene | AMPH1 |
| Simbolo del gene | AMPH |
| Specie | Wheat Germ (in vitro) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction