missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha Mouse anti-Human, Mouse, Rat, Clone: 1H8E1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66536-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Specifica
| AMPK alpha | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P54645, Q13131, Q5EG47 | |
| PRKAA1 | |
| AMPK alpha Fusion Protein Ag24744 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1H8E1 | |
| Unconjugated | |
| PRKAA1 | |
| AMPK, AMPK alpha, AMPK subunit alpha 1, AMPK1, AMPKa1, PRKAA1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 105787, 5562, 65248 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto