missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AmpSure™ Universal Probe qPCR Master Mix

Codice prodotto. 18972453 Sfoglia Tutto Applied Biosystems Prodotti
Cambia vista
Click to view available options
Quantità:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
18972453 5 mL
18962453 1 mL
18952453 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 18972453 Fornitore Thermo Scientific™ N. del fornitore A40013017
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Thermo Scientific™ AmpSure™ Universal Probe qPCR Master Mix is a ready-to-use, 2X concentrated master mix for probe-based qPCR detection and quantitation of a variety of DNA targets. Designed to support gene expression, copy number variation (CNV), single nucleotide polymorphism (SNP) genotyping, and microbial and pathogen detection.

Thermo Scientific™ AmpSure™ Universal Probe qPCR Master Mix is a ready-to-use, 2X concentrated master mix for probe-based qPCR detection and quantitation of a variety of DNA targets. Designed to support gene expression, copy number variation (CNV), single nucleotide polymorphism (SNP) genotyping, and microbial and pathogen detection. AmpSure Universal Probe qPCR Master Mix is a versatile master mix that can support multiple qPCR applications, instrument platforms, and suppliers' assays; simplifying reagent selection and laboratory workflows.

Features:

  • Competitive price per reaction for efficient qPCR workflows¹
  • Reliable multiplex gene expression analysis under fast and standard cycling conditions, helping to deliver PCR efficiencies up to 98% and R² values ≥0.999.
  • Precise SNP genotyping across fast and standard cycling protocols, designed to support up to 100% call rate and concordance results.
  • Accurate copy number variation analysis across multiple gene targets, helping to deliver up to 100% call rate.
  • Reliable analytical detection of pathogens and microbial DNA, designed to support ∣ΔCt∣ values as low as 0.01 and R² values up to 0.98.²
  • Performs on a wide range of leading qPCR instrument platforms
  • Compatible with TaqMan™ qPCR Assays and assays from other suppliers
  • Contains ROX™ passive reference dye for normalization on compatible instruments and heat-labile uracil-DNA glycosylase (UNG) to help reduce carryover contamination

Reliable Gene Expression Analysis

  • AmpSure Universal Probe qPCR Master Mix demonstrates sensitive and reliable gene expression analysis verified on several qPCR instrument platforms in fast and standard cycling protocols. The formulation is optimized for efficient amplification of multiplex (duplex) gene expression targets.

Precise SNP genotyping

  • AmpSure Universal Probe qPCR Master Mix demonstrates effective allelic discrimination in both fast and standard cycling conditions with both DME and non-DME assays. Using purified genomic DNA, the product helps deliver high call rate and accurate genotyping on a wide range of qPCR instrument platforms.

High accuracy CNV analysis

  • AmpSure Universal Probe qPCR Master Mix enables reliable copy number determination across multiple gene targets with accurate quantification in samples containing one to three copies. The product demonstrates consistent and accurate CNV analysis across a variety of qPCR instrument platforms.

Supports microbial and pathogen analytical detection

  • AmpSure Universal Probe qPCR Master Mix supports the analytical detection of microbial and pathogen DNA, with amplification of one or two targets across multiple qPCR platforms. Multiplex performance may require assay-specific optimization.

Compatible with multiple suppliers' assays

  • AmpSure Universal Probe qPCR Master Mix is compatible with a range of singleplex and duplex assay designs and chemistries. The universal formulation produces reliable results with both TaqMan and IDT™ assays when compared to IDT PrimeTime™ Gene Expression Master Mix in identical workflow configurations.

The master mix uses TaqMan fluorogenic 5' nuclease chemistry. During amplification, a target-specific probe containing reporter and quencher dyes anneals to the target sequence and is cleaved by the 5' nuclease activity of Taq DNA polymerase. Cleavage separates the reporter from the quencher, increasing fluorescence as amplification proceeds; the resulting fluorescence increase is proportional to the amount of amplicon produced.

AmpSure Universal Probe qPCR Master Mix is a versatile master mix that can help simplify protocol development and workflow integration, while reducing the potential for unused or expired application-specific reagents and incorrect master mix selection. Details of each of the qPCR application workflows, including recommended materials, sample preparation, reaction setup, and data analysis, are provided in the AmpSure Universal Probe qPCR Master Mix user guide included.

  1. Based on current list prices for comparable Applied Biosystems TaqMan qPCR Master Mixes.
  2. Multiplex performance may require assay-specific optimization.

TRUSTED_SUSTAINABILITY

Specifica

Concentrazione 2X (Master Mix)
Metodo di rilevamento qPCR
Forma Liquid
GC-Rich PCR Performance Medium
PCR Method 2-step RT-qPCR, qPCR, Multiplex qPCR
Reaction Speed Fast or Standard
Product Type qPCR Master Mix
Quantità 5 mL
Da utilizzare con (applicazione) Real Time PCR (qPCR), Gene Expression, Genotyping, Copy Number Variation Analysis, Pathogen Detection
N. di reazioni 500 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.