Learn More
IKA™ Frantoio a coltelli
GREENER_CHOICE
Codice prodotto. p-8007155 Sfoglia Tutto IKA Prodotti
Click to view available options
Modello:
A 10 Basic
A 11 Basic
Capacità (sistema metrico):
50 ml
80 ml
Durezza:
5 Mohs
6 Mohs
Dimensioni (L x P x A):
85 x 85 x 240 mm
145 x 130 x 250 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 10479412
Marca: IKA™ 0002900000
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Grinds hard, brittle, soft and fibrous materials. IKA™ Analytical Batch Mill allows samples to be embrittled directly in the grinding chamber, tough, oily and aqueous samples can also be ground. Available models include the newly redesigned A 10 Basic and the A 11 Basic.
Batch mill for two different grinding procedures. Impact grinding of hard, brittle or non-elastic grinding materials with high grade stainless steel beater. Beater (supplied) can be used for a Mohs hardness up to six. Cutting grinding for pulverising soft, fibrous materials with a cutting blade (must be ordered separately).
  • Moist and gluey materials can be pulverised by adding water
  • Grinding chamber (included with mill) made of ETFE (PTFE, glass fibre reinforced) with stainless steel inlet (A1S1 3/6L), with a useful volume of 80mL
  • Can be used for the embrittlement of grinding materials with liquid nitrogen in the grinding chamber
  • 250mL grinding chamber is available
TRUSTED_SUSTAINABILITY
Specifica
Product Type Frantoio a coltelli
Capacità (sistema metrico) 80 ml
Certificazione/Conformità DIN EN 60529 IP 43
Dimensioni (L x P x A) 85 x 85 x 240 mm
Requisiti elettrici 220/230/100/115 V, 50/60 Hz, 160 w
Durezza 6 Mohs
Altezza (sistema metrico) 240 mm
Hertz 50/60 Hz
Materiale Acciaio inox
Modello A 11 Basic
Dimensione particelle 10 mm
Tensione 220/230/100/115 V
Watt 160 w
Peso (sistema metrico) 1.5 kg
Larghezza (sistema metrico) 85 mm