Learn More
Abeba™ Anatom 2135 Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20345:2011 S2 SRC
- Microfiber black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area adjustable, with elastic band
- Suitable for heel and foot lift
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20345:2011 S2 SRC |
| Colore | Black |
| Materiale | Microfiber |
| Size | 48 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 2135 |
| Materiale dello zoccolo | Steel |