Learn More
Sampling Systems™ Angular Beaker
Codice prodotto. 17982032 Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Capacità (sistema metrico):
250 mL
500 mL
1 L
2 L
Materiale:
Polypropylene
Stainless Steel
Dimensione della confezione:
Pezzo
Codice prodotto. 17982032
Marca: Sampling Systems™ 5065B1001B
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
These angular beaker attachments allow the cup to be set in specific locations while using the TeleScoop.
  • Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
  • A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
  • Attachments are interchangeable and simply click into position
Specifica
Sterilità Non-sterile
Autoclavabile Autoclavable
Capacità (sistema metrico) 1 L
Monouso Reusable
Product Type Swabbing Template
Materiale Stainless Steel