missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD54 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 68388-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The ANKRD54 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ANKRD54. This antibody recognizes human, mouse, rat antigen. The ANKRD54 antibody has been validated for the following applications: WB, ELISA analysis.Specifica
| ANKRD54 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 129138 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, ELISA | |
| 1E9A9 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 33-35 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction