missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Antibody (ANXA1/3869R), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-08709PE
Questo articolo non è restituibile.
Consulta la politica di reso
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Spécification
| Annexin A1 | |
| Monoclonal | |
| PE | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Recombinant full-length human Annexin A1 protein (Uniprot: P04083) | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 301 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| ANXA1/3869R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu