Learn More
Annexin A2 Mouse anti-Human, CoraLite 594, Clone: 3H1B11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL594-60051-100UL
Descrizione
The Annexin A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Annexin A2. This antibody recognizes human, mouse antigen. The Annexin A2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifica
| Annexin A2 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 302 | |
| IgG2b |
| Immunofluorescence | |
| 3H1B11 | |
| Mouse | |
| Primary | |
| Human, Mouse |