Learn More
Annexin IV Mouse anti-Human, Clone: 1A10B5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60288-2-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.Specifica
| Annexin IV | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09525 | |
| ANXA4 | |
| Annexin IV Fusion Protein Ag0130 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2c |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1A10B5 | |
| Unconjugated | |
| ANXA4 | |
| 35-beta calcimedin, Annexin A4, Annexin IV, Annexin-4, Chromobindin-4, Endonexin I, Lipocortin IV | |
| Mouse | |
| Protein A | |
| RUO | |
| 307 | |
| -20°C | |
| Liquid |