Learn More
ATP-binding cassette, sub-family A (ABC1), member 6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ABCA6.
Marca: Abnova H00023460-A01.50uL
Descrizione
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This encoded protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. This gene is clustered among 4 other ABC1 family members on 17q24 and may play a role in macrophage lipid homeostasis. [provided by RefSeq
Sequence: NVQFPGMAPQNLGRVDKFNSSSLMVVYTPISNLTQQIMNKTALAPLLKGTSVIGAPNKTHMDEILLENLPYAMGIIFNETFSYKLIFFQGYNSPLWKSpecifica
| ATP-binding cassette, sub-family A (ABC1), member 6 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ABCA6. | |
| ABCA6 | |
| EST155051/FLJ43498 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 23460 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_080284 | |
| ABCA6 | |
| ABCA6 (NP_525023, 53 a.a. ∼ 149 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.