Learn More
ADCYAP1R1, Mouse, Clone: 2B12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ADCYAP1R1.
Marca: Abnova H00000117-M01.100ug
Descrizione
This gene encodes type I adenylate cyclase activating polypeptide receptor, which is a membrane-associated protein and shares significant homology with members of the glucagon/secretin receptor family. This receptor mediates diverse biological actions of adenylate cyclase activating polypeptide 1 and is positively coupled to adenylate cyclase. Alternative splicing of two exons of this gene generates four major splice variants, but their full-length nature has not been determined. [provided by RefSeq
Sequence: MHSDCIFKKEQAMCLEKIQRANELMGFNDSSPGCPGMWDNITCWKPAHVGEMVLVSCPELFRIFNPDQVWETETIGESDFGDSNSLDLSDMGVVSRNCTESpecifica
| ADCYAP1R1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001118 | |
| ADCYAP1R1 | |
| ADCYAP1R1 (NP_001109, 21 a.a. ∼ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2B12 | |
| Mouse monoclonal antibody raised against a partial recombinant ADCYAP1R1. | |
| ADCYAP1R1 | |
| PAC1/PACAPR/PACAPRI | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 117 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.