missing translation for 'onlineSavingsMsg'
Learn More

Beclin Mouse, Unlabeled, Clone: 20, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 612112

 vedi altre versioni di questo prodotto

Codice prodotto. 15826099

  • ONLINE EXCLUSIVE PRICE
    189.00€
    188.32€ / 1 set
    Risparmia 0.68€
    0.36% Sconto

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

Autophagy is the bulk degradation of cellular proteins through the autophagosomic-lysosomal pathway. This process is important for normal cell growth, and may be disrupted in tumor cells. Beclin is a Bcl-2 binding protein that is homologous to the yeast autophagy gene, apg6/vps30. The structure of beclin includes a Bcl-2 binding coiled-coil region, and a leucine-rich nuclear export signal (NES). Beclin protein colocalizes with intracytoplasmic organelles and nuclei in normal COS7 and MCF7 cells. However, inhibition of CRM1-dependent nuclear export leads to beclin localization primarily in the nucleus. Mutation of the NES domain of beclin also prevents nuclear export, as well as suppresses beclin-mediated nutrient deprivation-induced autophagy. In addition, these beclin mutants can not inhibit in vitro clonigenicity and in vivo tumorigenicity of MCF7 cells. Beclin interaction with Bcl-2 may be involved with host viral defense, since overexpression of beclin inhibits Sindbis virus replication and expression of beclin lacking the Bcl-2 binding domain has no antiviral effects. Thus, beclin may be an important component of complexes involved in autophagy.

Western Blotting

TRUSTED_SUSTAINABILITY
Specifica

Specifica

Beclin
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Beclin aa. 171-291
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG2a
Western Blot
20
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.