Learn More
CD3 Mouse anti-Human, APC-H7, Clone: SK7, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 560275
Descrizione
CD3 reacts with ε chain of the CD3 antigen/T-cell antigen receptor (TCR) complex. This complex is composed of at least six proteins that range in molecular weight from 20 to 30kDa. The antigen recognized by CD3 antibodies is noncovalently associated with either α/β or γ/δ TCR (70 to 90kDa). The CD3 antigen is present on 61% to 85% of normal peripheral blood lymphocytes 60% to 85% of thymocytes and on Purkinje cells in cerebellum. This antibody has a Mitogenic effect on most peripheral blood T lymphocytes, provided appropriate functional monocytes are present.
Host Species: Mouse
Clone: SK7
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifica
| CD3 | |
| Monoclonal | |
| APC-H7 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Human Thymocytes | |
| 25 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| SK7 | |
| CD3-epsilon; CD3E; Leu4; T-cell surface antigen T3/Leu-4 epsilon chain; T3E | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only.