Learn More
CD3 Mouse anti-Human, FITC, Clone: UCHT1, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 555332
Descrizione
The UCHT1 monoclonal antibody specifically binds to the human CD3ε-chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3ε-positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε.
Specifica
| CD3 | |
| Monoclonal | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| UCHT1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only.