Learn More
CD45 Rat anti-Mouse, Brilliant Violet 786, Clone: 30-F11, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 564225
Descrizione
The 30-F11 clone has been reported to react with all isoforms and both alloantigens of CD45, which is found on hematopoietic stem cells and all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane glycoprotein which is expressed at high levels on the cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family, where the intracellular carboxy-terminal region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated as A, B, and C, respectively). CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction and the CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifica
| CD45 | |
| Monoclonal | |
| 0.2mg/mL | |
| Ptprc; CD45R; LCA; Leukocyte common antigen; T200; B220; Ly-5; Lyt-4 | |
| Mouse Thymus / Spleen | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 30-F11 | |
| Brilliant Violet 786 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2b κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.