missing translation for 'onlineSavingsMsg'
Learn More

CD5 Rat anti-Mouse, PE, Clone: 53-7.3, BD

Rat Monoclonal Antibody

148.73€ - 737.23€

Specifica

Antigene CD5
Clone 53-7.3
Concentrazione 0.2mg/mL
Applicazioni Flow Cytometry
Classificazione Monoclonal
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
10095394
Vedere i documenti
BD Biosciences
553022
0.1 mg
ONLINE EXCLUSIVE PRICE
149.00€
148.73€ / Pezzo
Risparmia 0.27€
0.18% Sconto
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
15836938
Vedere i documenti
BD Biosciences
553023
0.2 mg
ONLINE EXCLUSIVE PRICE
740.00€
737.23€ / Pezzo
Risparmia 2.77€
0.37% Sconto
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

The 53-7.3 clone has been reported to react with a monomorphic epitope of CD5, a member of the scavenger receptor cysteine-rich protein superfamily and the major ligand of CD72, found on thymocytes, T lymphocytes, thymic NK-T cells, and a subset of B lymphocytes, but not on NK cells or splenic NK-T cells. The level of surface CD5 expression is developmentally regulated in the thymus, starting with low levels on CD4-CD8- thymocytes and increasing as they mature to CD4+CD8+ then CD4+CD8- or CD4-CD8+ thymocytes. Relatively high levels are maintained on peripheral T lymphocytes. The level of CD5 antigen detected on T helper cells has been reported to be somewhat higher than that on T cytotoxic/suppressor and B cells. Few, if any, intestinal intraepithelial lymphocytes bearing the γδ T-cell receptor express CD5. Phenotypic, anatomical, functional, developmental, and pathogenic characteristics of peripheral CD5+ B cells suggest that they may represent a distinct lineage, known as B-1 cells. The frequency of these CD5+ B cells has been reported to show strain-dependent variation. An additional population of CD5+ B lymphocytes resides in the thymus, where it matures from intrathymic B-cell progenitors. It has been proposed that CD5 is a costimulatory molecule which mediates interactions of cells in the immune system and negatively regulates signal transduction mediated by the T-cell receptor and B-cell receptor. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 53-7.3
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Thymus / Spleen

Flow Cytometry

Specifica

Specifica

CD5
0.2mg/mL
Monoclonal
Rat
Immunology
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
Lymphocyte antigen 1; Cd5; Ly-12; Ly-A; Lyt-1
53-7.3
Flow Cytometry
PE
RUO
Mouse
Mouse Thymus / Spleen
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documenti

Documenti

Product Certifications

For Research Use Only.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato