Learn More
CDC42 Mouse, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
180.83€ - 342.40€
Specifica
| Antigene | CDC42 |
|---|---|
| Clone | 44 |
| Concentrazione | 250μg/mL |
| Applicazioni | Western Blot |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
15865709
|
BD Biosciences
610928 |
50 μg |
181.00€ 180.83€ / 1 set Risparmia 0.17€ 0.09% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
15875709
|
BD Biosciences
610929 |
150 μg |
343.00€ 342.40€ / 1 set Risparmia 0.60€ 0.18% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Rho family members are small GTP binding proteins that serve as molecular switches for a number of biological processes. They cycle between active GTP-bound and inactive GDP-bound states. CDC42 is a Rho family protein that was identified in membranes of human platelets and placenta. It is the homologue of CDC42Sc, which regulates initiation of bud-site assembly in Saccharomyces cerevisiae. CDC42 regulates the function of the mammalian actin cytoskeleton, allowing for efficient cytokinesis and cell morphogenesis. CDC42 and Rac1, a Ras-related GTPase, activate MEKK1, a JNK kinase kinase, which leads to the activation of several downstream components of the MAP kinase cascade leading to activation of PAK65, a CDC42- and Rac-binding protein. PAK65 interacts with CDC42/Rac1, mediates their interaction with MEKK1, and enhances MEKK1 catalytic activity. Chronic activation of CDC42 has been shown to induce malignant cellular transformation. Due to sequence homology among various small GTPase family members, potential cross-reactivity could be observed with this antibody. BD Pharmingen has not performed such cross-reactivity testing.
Immunofluorescence, Western Blotting
Specifica
| CDC42 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 44 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Mouse, Rat | |
| Human CDC42 aa. 1-191 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.