Learn More
CCAAT/enhancer binding protein (C/EBP), gamma, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant CEBPG.
Marca: Abnova H00001054-A01.50uL
Descrizione
The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP alpha, C/EBP beta, C/EBP gamma, and C/EBP delta, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein gamma may cooperate with Fos to bind PRE-I enhancer elements. [provided by RefSeq
Sequence: MSKISQQNSTPGVNGISVIHTQAHASGLQQVPQLVPAGPGGGGKAVAPSKQSKKSSPMDRNSDEYRQRRERNNMAVKKSRLKSKQKAQDTLQRVNQLKEENERLEAKIKLLTKELSVLKDLFLEHAHNLADNVQSISTENTTADGDNAGQSpecifica
| CCAAT/enhancer binding protein (C/EBP), gamma | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant CEBPG. | |
| CEBPG | |
| GPE1BP/IG/EBP-1 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 1054 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC013128 | |
| CEBPG | |
| CEBPG (AAH13128, 1 a.a. ∼ 150 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.