missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clostridium difficile Toxin A Monoclonal Antibody (A73H), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA518192
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody does not cross-react with Toxin B. Purity is >90%. Recommended pairs for ELISA: MA5-18192 (capture) and MA5-18191 (conjugated, detection).
Clostridium difficile is a species of bacteria of the genus Clostridium which are Gram-positive, anaerobic, spore-forming rods (bacillus). C.difficile is the most significant cause of pseudomembranous colitis, a severe infection of the colon.Specifica
Clostridium difficile Toxin A | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA | |
A73H | |
Unconjugated | |
TcdA | |
Cl. difficile native protein | |
100 μg | |
Primary | |
4° C | |
Liquid |