Learn More
Haleco™ Anti-contamination Kit
Codice prodotto. 15447630
Click to view available options
Capacità (sistema metrico):
20 L
Dimensione della confezione:
Pezzo
Codice prodotto. 15447630
Marca: Haleco™ CKT0020SJ
Allows rapid intervention in small spills. Haleco™ Anit-contamination Kit includes flexible transport bag complete with 10 sheets, 2 sockets, 2 wipers, recovery bag with fasteners, and a pair of gloves.
Includes:
  • High visibility transport bag
  • 10 Sheets: 46 x 41cm
  • 2 Sockets: 8 x 122cm
  • 2 wipers
  • 1 recovery bag with fasteners
  • 1 pair of gloves
Specifica
Colore Yellow
Capacità (sistema metrico) 20 L
Tipo Spill Kit
Include High visibility transport bag
10 sheets: 46 x 41 cm
2 sockets: 8 x 122 cm
2 wipers
1 recovery bag with bandages 1 pair of gloves
Quantità confezione 1 Ea.
Peso (sistema metrico) 2 kg