Learn More
Cyclin A Mouse anti-Human, Unlabeled, Clone: 25, BD
Mouse Monoclonal Antibody
188.32€ - 383.06€
Specifica
| Antigene | Cyclin A |
|---|---|
| Clone | 25 |
| Concentrazione | 250μg/mL |
| Applicazioni | Western Blot |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
12820510
|
BD Biosciences
611268 |
50 μg |
189.00€ 188.32€ / Pezzo Risparmia 0.68€ 0.36% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
15835859
|
BD Biosciences
611269 |
150 μg |
384.00€ 383.06€ / 1 set Risparmia 0.94€ 0.25% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Progression of the mammalian cell cycle is regulated by phosphorylation of many key proteins. Several classes of cyclins (A-E) act as regulatory subunits for cyclin-dependent kinases (cdks). These cyclin-cdk holoenzymes are essential for proper control of cell cycle progression. They phosphorylate and regulate a variety of substrates whose activity is required for cell cycle transitions. The temporal expression of cyclins is tightly regulated throughout the cell cycle by synthesis and degradation. Such regulation plays a critical role in controlling the enzymatic activity of the cdks. Cyclin A, one of the mitotic cyclins, activates Cdk2 near the start of S phase and is necessary for the initiation of DNA replication. In mammalian somatic cells, Cyclin A is required during S phase and passage through G2. The D and E type cyclins regulate passage through G1, while Cyclin B is a critical regulator of mitosis. It has been shown in a number of species that mutation or disruption of normal Cyclin A expression causes cells to arrest at G2. Cyclin A binds both the cdc2 (Cdk1) and Cdk2 kinases and may also have a role in mitotic dependence on S phase completion.
Host Species: Mouse
Clone: 25
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Cyclin A aa. 26-144
Formula Weight [Chemical]: 60kDa
Bioimaging, Immunofluorescence, Western Blotting
Specifica
| Cyclin A | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 25 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human Cyclin A aa. 26-144 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.