Learn More
DKK3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA521325
Descrizione
PA5-21325 targets Dkk-3 in ICC, IF, and WB applications and shows reactivity with Human and mouse samples. The PA5-21325 immunogen is recombit protein fragment corresponding to a region within amino acids 6 and 343 (Q9UBP4) of DKK3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifica
| DKK3 | |
| Polyclonal | |
| Unconjugated | |
| Q9UBP4, Q9QUN9 | |
| DKK3 | |
| Recombit protein fragment corresponding to a region within amino acids 6 and 343 (Q9UBP4) of DKK3 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| DKK3 | |
| UNQ258/PRO295, REIC, RIG, RIG-like 5-6, RIG-like 7-1, dickkopf homolog 3, dickkopf-3, dickkopf-related protein 3, dkk-3, hDkk-3, regulated in glioma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27122, 50781 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.