Learn More
epidermal growth factor (beta-urogastrone), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EGF.
Marca: Abnova H00001950-A01.50uL
Descrizione
Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide. [provided by RefSeq
Sequence: NASCTNTEGGYTCMCAGRLSEPGLICPDSTPPPHLREDDHHYSVRNSDSECPLSHDGYCLHDGVCMYIEALDKYACNCVVGYIGERCQYRDLKWWKLRHASpecifica
| epidermal growth factor (beta-urogastrone) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EGF. | |
| EGF | |
| HOMG4/URG | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| 1950 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001963 | |
| EGF | |
| EGF (NP_001954, 926 a.a. ∼ 1025 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.