Learn More
FANCD2 Rabbit anti-Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 178 publications
Marca: Novus Biologicals NB100-182SS
Descrizione
FANCD2 Polyclonal specifically detects FANCD2 in Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, RNA Inhibition, Knockout Validated, Knockdown Validated.Specifica
| FANCD2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| Rabbit | |
| 164.1 kDa | |
| 0.025 mL | |
| Primary | |
| This antibody is specific for FANCD2. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, ChIP assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:2000, Simple Western 1:25, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 u | |
| Q9BXW9 | |
| FANCD2 | |
| This FANCD2 Antibody was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
| Affinity Purified | |
| RUO | |
| 2177 | |
| Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish | |
| IgG |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
For Research Use Only