Learn More
anti-FGF1 Matched Antibody Pair, Abnova
Marca: Abnova H00002246-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-FGF1 (100μg); Mouse purified polyclonal anti-FGF1 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis. Multiple alternatively spliced variants encoding different isoforms have been described.
ELISA
Specifica
| FGF1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FGF1. | |
| AFGF/ECGF/ECGF-beta/ECGFA/ECGFB/FGF-alpha/FGFA/GLIO703/HBGF1 | |
| Mouse, Rabbit | |
| 1 Set | |
| Angiogenesis | |
| 2246 | |
| Human |
| ELISA | |
| Unconjugated | |
| FGF1 | |
| FGF1 | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FGF1 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-FGF1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.