Learn More
Nitritex™ Anti-Fog Spray
Click to view available options
:
Quantity: 60mL
Descrizione
- Reduces fogging
- Airless pump action spray
- Non-hazardous
- Colorless
- Gamma-irradiated
- Alcohol free
- Quick and easy application
- Suitable for all BioClean autoclavable goggles
Specifica
Specifica
| Certificazione/Conformità | Manufactured in a facility holding ISO 9001:2008 certifications |
| Product Type | Anti-fog spray |
| Da utilizzare con (apparecchiatura) | Goggles |
| Da utilizzare con (applicazione) | Anti-fogging applications |
| Quantità | 60 mL |