Learn More
FYB Mouse anti-Human, Unlabeled, Clone: 5, BD
Mouse Monoclonal Antibody
Specifica
| Antigene | FYB |
|---|---|
| Clone | 5 |
| Concentrazione | 250μg/mL |
| Applicazioni | Immunofluorescence, Western Blot |
| Classificazione | Monoclonal |
Descrizione
Engagement of the T cell receptor (TcR) results in the activation of multiple intracellular src protein tyrosine kinases (PTKs) including p56 [lck] and p59 [fyn]. PTK activation induces TcRζ phosphorylation and the subsequent recruitment and activation of ZAP-70 which specifically phosphorylates SLP-76. SLP-76 interacts with multiple components of T cell signaling pathways, including Grb-2, PLC-γ, Vav and SLAP-130 (also known as SLP-76 Associated Phosphoprotein of 130kDa). SLAP-130 contains multiple tyrosine based motifs, proline base type I and II SH3 domain binding motifs, lysine/glutamic acid rich nuclear localization motifs, and an SH3-like domain. It associates with the SH2 domain of SLP-76 and is phosphorylated by TcR induced PTKs. This protein has also been reported to directly interact with fyn and has been referred to as fyn binding protein (FYB). Co-transfection data has shown that a limited concentration of FYB/SLAP-130 enhances TcR-induced IL-2 production, while overexpression downregulates gene expression. Thus, FYB/SLAP-130 is viewed to be a regulatory component of the fyn/SLP-76 signaling cascade in T cells.
Immunofluorescence, Western Blotting
Specifica
| FYB | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| Fyn Binding Protein; SLAP-130 |
| 5 | |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human FYB aa. 673-783 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.