missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCG, Human, Matched Antibody Pair (AP41), Abnova™
To detect and quantify protein level of human
Marca: Abnova H00002641-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-GCG, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-GCG, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifica
| GCG | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GCG. | |
| GLP1/GLP2/GRPP | |
| Mouse | |
| RUO | |
| 2641 | |
| Human | |
| Liquid |
| ELISA | |
| Biotin | |
| GCG | |
| GCG | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-GCG, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-GCG, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto